Assay Details
| Gene Symbol: Gm36011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030228.1 | - | 4 - 5 | 303 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA B130024G19 gene | B130024G19Rik | -683.8kb | Anti-sense | Chr. 7 - 70365383 - 70411146 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 6030442E23 gene | 6030442E23Rik | 121.6kb | Anti-sense | Chr. 7 - 71217049 - 71224410 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm36011
Genomic Map