Assay Details
| Gene Symbol: Gbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153564.2 | NP_705792.2 | 2 - 3 | 254 | 73 | |
| GenBank mRNA | AF422243.1 | - | 2 - 3 | 236 | 73 | |
| AK030414.1 | - | 2 - 3 | 254 | 73 | ||
| AK087083.1 | - | 342 | 73 | |||
| BC058555.1 | - | 2 - 3 | 288 | 5293575 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gbp5
Genomic Map