Assay Details
| Gene Symbol: Gbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153564.2 | NP_705792.2 | 6 - 7 | 940 | 79 | |
| GenBank mRNA | AF422243.1 | - | 6 - 7 | 922 | 79 | |
| AF487898.1 | - | 4 - 5 | 596 | 79 | ||
| AK030414.1 | - | 6 - 7 | 940 | 79 | ||
| AK087083.1 | - | 1028 | 79 | |||
| BC058555.1 | - | 6 - 7 | 974 | 5293575 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gbp5
Genomic Map