Assay Details
| Gene Symbol: Gbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153564.2 | NP_705792.2 | 7 - 8 | 1215 | 97 | |
| GenBank mRNA | AF422243.1 | - | 7 - 8 | 1197 | 97 | |
| AF487898.1 | - | 5 - 6 | 871 | 97 | ||
| AK030414.1 | - | 7 - 8 | 1215 | 97 | ||
| AK035481.1 | - | 1 - 2 | 2866 | 97 | ||
| AK087083.1 | - | 1303 | 97 | |||
| BC058555.1 | - | 7 - 8 | 1249 | 5293575 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gbp5
Genomic Map