Assay Details
| Gene Symbol: Gm29856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK082514.1 | - | 1 - 2 | 187 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger and BTB domain containing 18 | Zbtb18 |
RP58, Zfp238, Znf238 |
-8.2kb | Sense | Chr. 1 - 177442356 - 177450764 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 2310043L19 gene | 2310043L19Rik | 181.3kb | Anti-sense | Chr. 1 - 177641542 - 177642943 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm29856
Genomic Map