Assay Details
| Gene Symbol: Unc45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133952.2 | NP_598713.2 | 18 - 19 | 2524 | 63 | |
| GenBank mRNA | AK028912.1 | - | 18 - 19 | 2510 | 63 | |
| AK047292.1 | - | 18 - 19 | 2471 | 63 | ||
| AK047476.1 | - | 18 - 19 | 2471 | 63 | ||
| AK088034.1 | - | 18 - 19 | 2445 | 63 | ||
| AK166809.1 | - | 18 - 19 | 2507 | 63 | ||
| AK203626.1 | - | 1 - 2 | 63 | |||
| BC004717.1 | - | 18 - 19 | 2513 | 3582116 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Unc45a
Genomic Map