Assay Details
| Gene Symbol: Alpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027808.1 | NP_082084.1 | 2 - 3 | 603 | 98 | |
| GenBank mRNA | AK018401.1 | - | 2 - 3 | 573 | 98 | |
| AK041315.1 | - | 2 - 3 | 590 | 98 | ||
| AK173191.1 | - | 2 - 3 | 603 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alpk1
Genomic Map