Assay Details
| Gene Symbol: Alpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027808.1 | NP_082084.1 | 4 - 5 | 951 | 61 | |
| GenBank mRNA | AK173191.1 | - | 4 - 5 | 951 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alpk1
Genomic Map