Assay Details
| Gene Symbol: Ak5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081277.1 | NP_001074746.1 | 2 - 3 | 532 | 65 | |
| GenBank mRNA | AK053807.1 | - | 2 - 3 | 532 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ak5
Genomic Map