Assay Details
| Gene Symbol: Cblc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023224.5 | NP_075713.3 | 6 - 7 | 1027 | 80 | |
| GenBank mRNA | AF319956.1 | - | 6 - 7 | 1000 | 80 | |
| AK010228.1 | - | 6 - 7 | 1028 | 80 | ||
| BC046337.1 | - | 6 - 7 | 1003 | 6395806 | 80 | |
| BE198430.1 | - | 4 - 5 | 370 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cblc
Genomic Map