Assay Details
| Gene Symbol: Cblc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023224.5 | NP_075713.3 | 7 - 8 | 1160 | 91 | |
| GenBank mRNA | AF319956.1 | - | 7 - 8 | 1133 | 91 | |
| AK010228.1 | - | 7 - 8 | 91 | |||
| BC046337.1 | - | 7 - 8 | 1136 | 6395806 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cblc
Genomic Map