Assay Details
| Gene Symbol: Ccbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293560.1 | NP_001280489.1 | 2 - 3 | 231 | 107 | |
| GenBank mRNA | AF363737.1 | - | 2 - 3 | 205 | 107 | |
| AK145623.1 | - | 2 - 3 | 229 | 107 | ||
| BC085287.1 | - | 2 - 3 | 186 | 107 | ||
More Information
Set Membership: |
|

Ccbl2
Genomic Map