Assay Details
| Gene Symbol: Ccbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293560.1 | NP_001280489.1 | 3 - 4 | 287 | 104 | |
| GenBank mRNA | AF363737.1 | - | 3 - 4 | 261 | 104 | |
| AK145623.1 | - | 3 - 4 | 285 | 104 | ||
| BC085287.1 | - | 3 - 4 | 242 | 104 | ||
| BC131942.1 | - | 2 - 3 | 76 | 40130219 | 104 | |
| BC132615.1 | - | 2 - 3 | 76 | 40130892 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC131942.1, BC132615.1 transcripts.
Set Membership: |
|

Ccbl2
Genomic Map