Assay Details
| Gene Symbol: 4931440J10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045503.1 | 2 - 3 | 163 | 157 | ||
| GenBank mRNA | AK016511.1 | - | 2 - 3 | 164 | 157 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | integrator complex subunit 6 | Ints6 |
2900075H24Rik, AI480962, DICE1, Ddx26, HDB, Notch2l |
-4.6kb | Anti-sense | Chr. 14 - 62676325 - 62761370 on NCBI Build GRCm38 |
| Downstream | WD repeat and FYVE domain containing 2 | Wdfy2 |
5830485M08, B130024L21Rik, ProF, ZFYVE22 |
24.8kb | Sense | Chr. 14 - 62837690 - 62961509 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4931440J10Rik
Genomic Map