Assay Details
| Gene Symbol: Zdhhc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172465.4 | NP_766053.1 | 3 - 4 | 673 | 156 | |
| GenBank mRNA | AK032233.1 | - | 3 - 4 | 655 | 156 | |
| AK035745.1 | - | 2 - 3 | 2096 | 156 | ||
| AK079248.1 | - | 3 - 4 | 640 | 157 | ||
| BC042618.1 | - | 3 - 4 | 532 | 4950217 | 156 | |
| BC090832.1 | - | 3 - 4 | 491 | 6401763 | 156 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc9
Genomic Map