Assay Details
| Gene Symbol: Slc7a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253679.1 | NP_001240608.1 | 6 - 7 | 94 | ||
| NM_001253680.1 | NP_001240609.1 | 5 - 6 | 94 | |||
| NM_011405.4 | NP_035535.2 | 7 - 8 | 94 | |||
| GenBank mRNA | AJ012754.1 | - | 5 - 6 | 1155 | 94 | |
| AJ130943.1 | - | 6 - 7 | 1147 | 94 | ||
| AK150950.1 | - | 7 - 8 | 94 | |||
| AK151718.1 | - | 7 - 8 | 94 | |||
| AK152637.1 | - | 5 - 6 | 94 | |||
| AK170851.1 | - | 5 - 6 | 1076 | 94 | ||
| BC014709.1 | - | 5 - 6 | 1153 | 4482816 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a7
Genomic Map