Assay Details
| Gene Symbol: Hdgfrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013886.4 | NP_038914.2 | 3 - 4 | 737 | 114 | |
| GenBank mRNA | AB029493.1 | - | 3 - 4 | 300 | 114 | |
| AK019487.1 | - | 3 - 4 | 400 | 114 | ||
| AK044858.1 | - | 3 - 4 | 562 | 114 | ||
| AK045542.1 | - | 3 - 4 | 737 | 114 | ||
| AK051401.1 | - | 3 - 4 | 659 | 114 | ||
| AK162418.1 | - | 3 - 4 | 380 | 114 | ||
| AK162967.1 | - | 3 - 4 | 301 | 114 | ||
| BC055734.1 | - | 3 - 4 | 324 | 6810355 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hdgfrp3
Genomic Map