Assay Details
| Gene Symbol: 2900076A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045299.1 | 3 - 4 | 384 | 102 | ||
| GenBank mRNA | AK013794.1 | - | 2 - 3 | 250 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cDNA sequence BC048679 | BC048679 | -30.8kb | Anti-sense | Chr. 7 - 81494272 - 81498330 on NCBI Build GRCm38 | |
| Downstream | microRNA 1839 | Mir1839 |
mir-1839, mmu-mir-1839 |
389.0 | Sense | Chr. 7 - 81529916 - 81529988 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

2900076A07Rik
Genomic Map