Assay Details
| Gene Symbol: 2700070H01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046019.1 | 1 - 2 | 31 | 73 | ||
| GenBank mRNA | AK012509.1 | - | 1 - 2 | 31 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | defensin beta 42 | Defb42 | Defb44 | -6.5kb | Sense | Chr. 14 - 63046991 - 63048606 on NCBI Build GRCm38 |
| Downstream | cathepsin B | Ctsb | CB | 66.5kb | Sense | Chr. 14 - 63122462 - 63145923 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

2700070H01Rik
Genomic Map