Assay Details
| Gene Symbol: Ceacam20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027839.2 | NP_082115.2 | 3 - 4 | 487 | 79 | |
| GenBank mRNA | AK018613.1 | - | 3 - 4 | 484 | 79 | |
| AK033581.2 | - | 3 - 4 | 488 | 79 | ||
| BC049361.1 | - | 3 - 4 | 468 | 6476373 | 79 | |
More Information
Set Membership: |
|

Ceacam20
Genomic Map