Assay Details
| Gene Symbol: Msh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282054.1 | NP_001268983.1 | 10 - 11 | 1434 | 138 | |
| NM_031870.3 | NP_114076.1 | 9 - 10 | 1525 | 138 | ||
| GenBank mRNA | AF178957.1 | - | 9 - 10 | 1229 | 138 | |
| AF298655.1 | - | 9 - 10 | 1525 | 138 | ||
| AK029837.1 | - | 10 - 11 | 1435 | 138 | ||
| AK161474.1 | - | 9 - 10 | 1275 | 138 | ||
| AY351585.1 | - | 9 - 10 | 1479 | 138 | ||
| AY351590.1 | - | 1 - 2 | 866 | 138 | ||
| AY351591.1 | - | 1 - 2 | 866 | 138 | ||
| BC138163.1 | - | 9 - 10 | 1416 | 8861182 | 138 | |
| BC145838.1 | - | 9 - 10 | 1416 | 40131141 | 138 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY351590.1, AY351591.1 transcripts.
Set Membership: |
|

Msh4
Genomic Map