Assay Details
| Gene Symbol: Olfm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK133536.1 | - | 4 - 5 | 731 | 75 | |
| BC141127.1 | - | 4 - 5 | 780 | 9055739 | 75 | |
| BC145436.1 | - | 4 - 5 | 780 | 9053994 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Olfm4
Genomic Map