Assay Details
| Gene Symbol: Ppa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146141.2 | NP_666253.1 | 11 - 12 | 989 | 73 | |
| GenBank mRNA | AK003660.1 | - | 6 - 7 | 1274 | 73 | |
| AK143475.1 | - | 11 - 12 | 960 | 73 | ||
| BC011417.1 | - | 11 - 12 | 983 | 2654305 | 73 | |
More Information
Set Membership: |
|

Ppa2
Genomic Map