Assay Details
| Gene Symbol: Alpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027808.1 | NP_082084.1 | 11 - 12 | 3623 | 68 | |
| GenBank mRNA | AK173191.1 | - | 10 - 11 | 5335 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alpk1
Genomic Map