Assay Details
| Gene Symbol: Alpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027808.1 | NP_082084.1 | 8 - 9 | 1268 | 101 | |
| GenBank mRNA | AK173191.1 | - | 7 - 8 | 2980 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alpk1
Genomic Map