Assay Details
| Gene Symbol: Alpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027808.1 | NP_082084.1 | 9 - 10 | 81 | ||
| GenBank mRNA | AK173191.1 | - | 8 - 9 | 3094 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alpk1
Genomic Map