Assay Details
| Gene Symbol: Wls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026582.4 | NP_080858.3 | 7 - 8 | 1311 | 100 | |
| NR_037590.1 | 8 - 9 | 1392 | 100 | |||
| GenBank mRNA | AK019886.1 | - | 8 - 9 | 1350 | 100 | |
| AK029053.1 | - | 7 - 8 | 1270 | 100 | ||
| AK029104.1 | - | 7 - 8 | 1268 | 100 | ||
| AK029315.1 | - | 7 - 8 | 1268 | 100 | ||
| AK044810.1 | - | 7 - 8 | 1270 | 100 | ||
| AK049842.1 | - | 7 - 8 | 1268 | 100 | ||
| AK077407.1 | - | 7 - 8 | 1269 | 100 | ||
| AK128912.1 | - | 7 - 8 | 1267 | 100 | ||
| AK159207.1 | - | 7 - 8 | 1268 | 100 | ||
| AK159977.1 | - | 7 - 8 | 1270 | 100 | ||
| BC018381.1 | - | 6 - 7 | 895 | 4237619 | 100 | |
| BC075615.1 | - | 7 - 8 | 1311 | 5703610 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wls
Genomic Map