Assay Details
| Gene Symbol: Dtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB095735.1 | - | 6 - 7 | 647 | 109 | |
| AB095736.1 | - | 6 - 7 | 647 | 109 | ||
| AK012919.1 | - | 6 - 7 | 648 | 109 | ||
| AK054412.1 | - | 6 - 7 | 637 | 109 | ||
| AK133177.1 | - | 6 - 7 | 646 | 109 | ||
| AK154594.1 | - | 7 - 8 | 687 | 109 | ||
| AK161401.1 | - | 3 - 4 | 248 | 109 | ||
| AK164249.1 | - | 6 - 7 | 647 | 109 | ||
| AK164999.1 | - | 6 - 7 | 646 | 109 | ||
| AK166351.1 | - | 6 - 7 | 655 | 109 | ||
| AK169601.1 | - | 6 - 7 | 646 | 109 | ||
More Information
Set Membership: |
|

Dtl
Genomic Map