Assay Details
| Gene Symbol: Ppp2r5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144880.4 | NP_659129.2 | 9 - 10 | 1606 | 152 | |
| GenBank mRNA | AK145397.1 | - | 10 - 11 | 992 | 152 | |
| BC023062.1 | - | 3 - 4 | 176 | 152 | ||
| BC059026.1 | - | 9 - 10 | 1606 | 6810754 | 152 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC023062.1 transcripts.
Set Membership: |
|

Ppp2r5a
Genomic Map