| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100031057.mm.1 |
NM_001081259.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031057.mm.1 |
NM_001081259.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031057.mm.1 |
TC0100003797.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100031058.mm.1 |
NM_001081259.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031058.mm.1 |
NM_001081259.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031058.mm.1 |
TC0100003797.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100031061.mm.1 |
NM_001081259.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031061.mm.1 |
NM_001081259.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031061.mm.1 |
TC0100003797.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100031062.mm.1 |
NM_001081259.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031062.mm.1 |
NM_001081259.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031062.mm.1 |
TC0100003797.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100031060.mm.1 |
NM_001081259.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031060.mm.1 |
NM_001081259.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031060.mm.1 |
TC0100003797.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100031059.mm.1 |
NM_001081259.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031059.mm.1 |
NM_001081259.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031059.mm.1 |
TC0100003797.mm.1 |
MTA TC |