Assay Details
| Gene Symbol: Mfsd7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081259.1 | NP_001074728.1 | 4 - 5 | 1208 | 114 | |
| GenBank mRNA | AK036315.1 | - | 3 - 4 | 805 | 114 | |
| AK165872.1 | - | 4 - 5 | 1208 | 114 | ||
| BC157992.1 | - | 4 - 5 | 1176 | 9007305 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd7b
Genomic Map