Assay Details
| Gene Symbol: Mfsd7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081259.1 | NP_001074728.1 | 5 - 6 | 1310 | 76 | |
| GenBank mRNA | AK036315.1 | - | 4 - 5 | 907 | 76 | |
| AK165872.1 | - | 5 - 6 | 1310 | 76 | ||
| BC157992.1 | - | 5 - 6 | 1278 | 9007305 | 76 | |
More Information
Set Membership: |
|

Mfsd7b
Genomic Map