Assay Details
| Gene Symbol: Mfsd7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081259.1 | NP_001074728.1 | 6 - 7 | 1414 | 68 | |
| GenBank mRNA | AK036315.1 | - | 5 - 6 | 1011 | 68 | |
| AK165872.1 | - | 6 - 7 | 1414 | 68 | ||
| BC024751.1 | - | 1 - 2 | 392 | 68 | ||
| BC157992.1 | - | 6 - 7 | 1382 | 9007305 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd7b
Genomic Map