Assay Details
| Gene Symbol: Nsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198654.3 | NP_941056.3 | 4 - 5 | 515 | 64 | |
| GenBank mRNA | BC029086.1 | - | 4 - 5 | 505 | 3156934 | 64 |
More Information
Updated Mapping Information
Set Membership: |
|

Nsl1
Genomic Map