Assay Details
| Gene Symbol: Metap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175224.4 | NP_780433.1 | 5 - 6 | 445 | 112 | |
| GenBank mRNA | AK129054.1 | - | 1 - 2 | 938 | 112 | |
| AK152138.1 | - | 5 - 6 | 464 | 112 | ||
| AK153231.1 | - | 5 - 6 | 464 | 112 | ||
| AK153944.1 | - | 5 - 6 | 453 | 112 | ||
| BC096469.1 | - | 5 - 6 | 436 | 3603051 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Metap1
Genomic Map