Assay Details
| Gene Symbol: Metap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175224.4 | NP_780433.1 | 6 - 7 | 531 | 103 | |
| GenBank mRNA | AK077694.1 | - | 6 - 7 | 532 | 103 | |
| AK152138.1 | - | 6 - 7 | 550 | 103 | ||
| AK153231.1 | - | 6 - 7 | 550 | 103 | ||
| AK153944.1 | - | 6 - 7 | 539 | 103 | ||
| BC027110.1 | - | 1 - 2 | 71 | 103 | ||
| BC096469.1 | - | 6 - 7 | 522 | 3603051 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Metap1
Genomic Map