Assay Details
| Gene Symbol: Fabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007980.3 | NP_032006.1 | 3 - 4 | 685 | 66 | |
| GenBank mRNA | AK007995.1 | - | 3 - 4 | 383 | 66 | |
| AY523818.1 | - | 3 - 4 | 371 | 66 | ||
| AY523819.1 | - | 3 - 4 | 371 | 66 | ||
| BC013457.1 | - | 3 - 4 | 365 | 4218050 | 66 | |
| BY707992.1 | - | 3 - 4 | 381 | 66 | ||
| M65034.1 | - | 3 - 4 | 379 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P313581 | BC013457.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P313581 | NM_007980.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P313581 | AK007995.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P313581 | M65034.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P313581 | BC013457.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P313581 | NM_007980.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P313581 | AK007995.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P313581 | M65034.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 35286 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780280 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 753892 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 42670 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108151 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P313581 | BC013457.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P313581 | NM_007980.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P313581 | AK007995.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P313581 | M65034.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P313581 | BC013457.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P313581 | NM_007980.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P313581 | AK007995.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P313581 | M65034.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P313581 | BC013457.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P313581 | NM_007980.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P313581 | AK007995.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P313581 | M65034.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P313581 | BC013457.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P313581 | NM_007980.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P313581 | AK007995.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P313581 | M65034.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22485095 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108151 | - | - |
| Whole Mouse Genome G4122A | A_51_P313581 | BC013457.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P313581 | NM_007980.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P313581 | AK007995.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P313581 | M65034.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P313581 | BC013457.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P313581 | NM_007980.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P313581 | AK007995.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P313581 | M65034.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P453013 | AY523819.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P453013 | BC013457.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P453013 | NM_007980.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P453013 | AK007995.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P453013 | AY523818.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108703 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108151 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02815820 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02861055 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02826367 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02826362 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02815818 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02826363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22485137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02763086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02826377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02303550 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02826368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02826367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22485084 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10495823 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495825 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495824 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495822 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495821 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418438_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10495820 | - | - |
| Mouse Genome 430 2.0 Array | 1418438_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418438_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14079_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14079_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300010230.mm.1 | NM_007980.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300010230.mm.1 | TC0300001251.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300010227.mm.1 | NM_007980.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300010227.mm.1 | TC0300001251.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300010228.mm.1 | NM_007980.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300010228.mm.1 | TC0300001251.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300010229.mm.1 | NM_007980.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300010229.mm.1 | TC0300001251.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300010231.mm.1 | NM_007980.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300010231.mm.1 | TC0300001251.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418438_at | - | - |
| Mouse Expression Set 430 | 1418438_PM_at | - | - |
| Murine Genome U74v2 Set | 97889_at | - | - |
| Murine Genome U74v2 Set | 97889_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418438_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418438_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418438_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418438_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 859800 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5050673 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005050673 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2706493 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2706493 | - | - |
| mouse-6 v1.1 (Illumina) | scl22660.4.1_106-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5050673 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22660.4.1_106-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5050673 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2706493 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2706493 | - | - |

Fabp2
Genomic Map