Assay Details
| Gene Symbol: Mesp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008589.2 | NP_032615.2 | 1 - 2 | 1034 | 66 | |
| GenBank mRNA | AK146740.1 | - | 1 - 2 | 514 | 66 | |
| BC145652.1 | - | 1 - 2 | 864 | 40130955 | 66 | |
| BC145654.1 | - | 1 - 2 | 864 | 40130957 | 66 | |
More Information
Set Membership: |
|

Mesp2
Genomic Map