Assay Details
| Gene Symbol: Sema4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013659.4 | NP_038687.2 | 2 - 3 | 407 | 81 | |
| GenBank mRNA | AK028896.1 | - | 2 - 3 | 370 | 81 | |
| AK134973.1 | - | 1 - 2 | 335 | 81 | ||
| AK145570.1 | - | 2 - 3 | 408 | 81 | ||
| AK154969.1 | - | 1 - 2 | 188 | 81 | ||
| AK156620.1 | - | 1 - 2 | 241 | 81 | ||
| AK173249.1 | - | 2 - 3 | 355 | 81 | ||
| DQ073331.1 | - | 1 - 2 | 163 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema4b
Genomic Map