Assay Details
| Gene Symbol: Pdha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008811.2 | NP_032837.1 | 1 | 2307 | 68 | |
| GenBank mRNA | M76728.1 | - | 1 | 2304 | 68 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
glucose metabolic process
acetyl-CoA biosynthetic process from pyruvate
glycolytic process
tricarboxylic acid cycle
metabolic process
oxidation-reduction process
pyruvate dehydrogenase (acetyl-transferring) activity
oxidoreductase activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, disulfide as acceptor
carbohydrate metabolic process
glucose metabolic process
acetyl-CoA biosynthetic process from pyruvate
glycolytic process
tricarboxylic acid cycle
metabolic process
oxidation-reduction process
pyruvate dehydrogenase (acetyl-transferring) activity
oxidoreductase activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, disulfide as acceptor
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22582958 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22582961 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22582957 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22582960 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22582962 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22582963 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125079 | M76728.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125079 | NM_008811.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125079 | AK006356.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125079 | AK076791.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125079 | BC100460.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171353 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125079 | M76728.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125079 | NM_008811.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125079 | AK006356.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125079 | AK076791.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125079 | BC100460.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519857 | M76728.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519857 | NM_008811.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519857 | AK006356.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519857 | AK076791.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519857 | BC100460.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22583001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22583011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22583005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22583006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22583002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22583003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22583007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22582950 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171353 | - | - |
| Whole Mouse Genome G4122A | A_51_P519857 | M76728.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519857 | NM_008811.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519857 | AK006356.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519857 | AK076791.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519857 | BC100460.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125079 | M76728.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125079 | NM_008811.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125079 | AK006356.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125079 | AK076791.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125079 | BC100460.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171353 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02672053 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02977132 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02977127 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02672052 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02977135 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02977133 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02672055 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22582963 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22582961 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22582960 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22582958 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22582962 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22582957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22582933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22583025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583018 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02600385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02774016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02977132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582997 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02977142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02774010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582938 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02600394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03166675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02551878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02600400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03142077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02551879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02551883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02600392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02600369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22583003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02977127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22582991 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10502448 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10502449 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10502450 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10502447 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450962_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10502446 | - | - |
| Mouse Genome 430 2.0 Array | 1450962_at | - | - |
| Mouse Genome 430 2.0 Array | 1450962_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18598_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18598_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa13440_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300023919.mm.1 | NM_008811.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300023919.mm.1 | NM_008811.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300023919.mm.1 | TC0300003036.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450962_PM_at | - | - |
| Mouse Expression Set 430 | 1450962_at | - | - |
| Murine Genome U74v2 Set | 99542_PM_at | - | - |
| Murine Genome U74v2 Set | 99542_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450962_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450962_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450962_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450962_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 749653 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2630438 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002630438 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2667428 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2667428 | - | - |
| mouse-6 v1.1 (Illumina) | scl21451.1.529_18-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2630438 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21451.1.529_18-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2630438 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2667428 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2667428 | - | - |

Pdha2
Genomic Map