Assay Details
| Gene Symbol: Adss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007422.3 | NP_031448.2 | 10 - 11 | 1181 | 154 | |
| GenBank mRNA | AK004877.1 | - | 10 - 11 | 1134 | 154 | |
| AK010263.1 | - | 10 - 11 | 1156 | 154 | ||
| AK028060.1 | - | 10 - 11 | 1181 | 154 | ||
| AK148420.1 | - | 10 - 11 | 1163 | 154 | ||
| AK170279.1 | - | 10 - 11 | 1167 | 154 | ||
| BC139404.1 | - | 10 - 11 | 1093 | 8862426 | 154 | |
| BC139417.1 | - | 10 - 11 | 1093 | 8862439 | 154 | |
| L24554.1 | - | 10 - 11 | 1148 | 154 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adss
Genomic Map