Assay Details
| Gene Symbol: Adss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007422.3 | NP_031448.2 | 5 - 6 | 592 | 69 | |
| GenBank mRNA | AK004877.1 | - | 5 - 6 | 545 | 69 | |
| AK010263.1 | - | 5 - 6 | 567 | 69 | ||
| AK028060.1 | - | 5 - 6 | 592 | 69 | ||
| AK043273.1 | - | 5 - 6 | 585 | 69 | ||
| AK148420.1 | - | 5 - 6 | 574 | 69 | ||
| AK170279.1 | - | 5 - 6 | 578 | 69 | ||
| BC139404.1 | - | 5 - 6 | 504 | 8862426 | 69 | |
| BC139417.1 | - | 5 - 6 | 504 | 8862439 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adss
Genomic Map