Assay Details
| Gene Symbol: Adss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007422.3 | NP_031448.2 | 8 - 9 | 905 | 80 | |
| GenBank mRNA | AK004877.1 | - | 8 - 9 | 858 | 80 | |
| AK010263.1 | - | 8 - 9 | 880 | 80 | ||
| AK028060.1 | - | 8 - 9 | 905 | 80 | ||
| AK148420.1 | - | 8 - 9 | 887 | 80 | ||
| AK170279.1 | - | 8 - 9 | 891 | 80 | ||
| BC139404.1 | - | 8 - 9 | 817 | 8862426 | 80 | |
| BC139417.1 | - | 8 - 9 | 817 | 8862439 | 80 | |
| L24554.1 | - | 8 - 9 | 872 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adss
Genomic Map