Assay Details
| Gene Symbol: Ifi44l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031367.1 | NP_112735.1 | 1130 | 156 | ||
| GenBank mRNA | AF336221.1 | - | 1130 | 156 | ||
| BC024930.1 | - | 1001 | 4020690 | 156 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifi44l
Genomic Map