Assay Details
| Gene Symbol: Ndst4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022565.2 | NP_072087.2 | 10 - 11 | 2776 | 57 | |
| GenBank mRNA | AB036838.1 | - | 9 - 10 | 2226 | 57 | |
| AK132938.1 | - | 5 - 6 | 931 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndst4
Genomic Map