Assay Details
| Gene Symbol: Ugt8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011674.4 | NP_035804.2 | 3 - 4 | 1059 | 96 | |
| GenBank mRNA | AK128994.1 | - | 3 - 4 | 1055 | 96 | |
| AK137364.1 | - | 3 - 4 | 1060 | 96 | ||
| AK144005.1 | - | 3 - 4 | 1059 | 96 | ||
| AK163650.1 | - | 3 - 4 | 1021 | 96 | ||
| BC016885.1 | - | 1054 | 4223057 | 96 | ||
| X92122.1 | - | 2 - 3 | 1384 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ugt8a
Genomic Map