Assay Details
| Gene Symbol: Pycr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133705.2 | NP_598466.1 | 3 - 4 | 511 | 106 | |
| GenBank mRNA | AK007535.1 | - | 3 - 4 | 492 | 106 | |
| AK165618.1 | - | 3 - 4 | 486 | 106 | ||
| AK181506.1 | - | 1 - 2 | 67 | 106 | ||
| AK184426.1 | - | 1 - 2 | 67 | 106 | ||
| BC006882.1 | - | 3 - 4 | 487 | 2647612 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pycr2
Genomic Map