Assay Details
| Gene Symbol: Acbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133225.3 | NP_573488.2 | 1 - 2 | 329 | 102 | |
| GenBank mRNA | AF022770.3 | - | 1 - 2 | 330 | 102 | |
| AK030371.1 | - | 1 - 2 | 330 | 102 | ||
| AK167272.1 | - | 1 - 2 | 311 | 102 | ||
| BC050121.1 | - | 1 - 2 | 300 | 102 | ||
| BC060602.1 | - | 1 - 2 | 284 | 5706182 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acbd3
Genomic Map