Assay Details
| Gene Symbol: Parp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007415.2 | NP_031441.2 | 10 - 11 | 1601 | 89 | |
| GenBank mRNA | AK028042.1 | - | 10 - 11 | 1620 | 89 | |
| AK030889.1 | - | 10 - 11 | 1604 | 89 | ||
| AK088679.1 | - | 10 - 11 | 1605 | 89 | ||
| AK146677.1 | - | 10 - 11 | 1622 | 89 | ||
| AK152360.1 | - | 10 - 11 | 1622 | 89 | ||
| AK165158.1 | - | 10 - 11 | 1621 | 89 | ||
| AK166104.1 | - | 10 - 11 | 1653 | 89 | ||
| AK167699.1 | - | 10 - 11 | 1593 | 89 | ||
| AK169326.1 | - | 10 - 11 | 1622 | 89 | ||
| BC012041.1 | - | 10 - 11 | 1601 | 2648390 | 89 | |
| X14206.1 | - | 10 - 11 | 89 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Parp1
Genomic Map