Assay Details
| Gene Symbol: Tmem63a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144794.2 | NP_659043.1 | 12 - 13 | 1137 | 77 | |
| GenBank mRNA | AK049360.1 | - | 12 - 13 | 1131 | 77 | |
| AK144899.1 | - | 12 - 13 | 1190 | 77 | ||
| AK170559.1 | - | 12 - 13 | 1153 | 77 | ||
| AK173027.1 | - | 12 - 13 | 1130 | 77 | ||
| BC014795.1 | - | 12 - 13 | 1110 | 3961992 | 77 | |
| BC019442.1 | - | 12 - 13 | 1137 | 4036849 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem63a
Genomic Map